2-(4-tert-Butylphenyl)-5-(4-biphenylyl)-1,3,4-oxadiazole ≥98.0% (by HPLC)

Supplier: TCI America

B1767-5G
TCB1767-005GEA 139.04 CAD
TCB1767-005G
2-(4-tert-Butylphenyl)-5-(4-biphenylyl)-1,3,4-oxadiazole ≥98.0% (by HPLC)
2-(4-tert-Butylphenyl)-5-(4-biphenylyl)-1,3,4-oxadiazole

CAS Number: 15082-28-7
MDL Number: MFCD00003101
Molecular Formula: C24H22N2O
Molecular Weight: 354.45
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 137


Product Categories: Materials Chemistry, Organic Light-Emitting Diode (OLED) Materials, Analytical Science, Scintillators
Other Product Names / Synonyms: 2-(4-Biphenylyl)-5-(4-tert-butylphenyl)-1,3,4-oxadiazole, PBD

Formula: C₂₄H₂₂N₂O
Melting Pt: 137 °C
MDL Number: MFCD00003101
CAS Number: 15082-28-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5cf7a4fd9e508b3e4943be56afb6dbc190350341&vid=5cf7a4fd9e508b3e4943be56afb6dbc190350341&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR