3,5-Bis(trifluoromethyl)benzyl alcohol ≥98.0%

Supplier: TCI America

Synonyms: (3,5-Bis(trifluoromethyl)phenyl)methanol

B1752-5G
TCB1752-005GEA 116.92 CAD
TCB1752-005G
3,5-Bis(trifluoromethyl)benzyl alcohol ≥98.0%
3,5-Bis(trifluoromethyl)benzyl alcohol

CAS Number: 32707-89-4
MDL Number: MFCD00009907
Molecular Formula: C9H6F6O
Molecular Weight: 244.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 55
Flash Point (°C): 98

Formula: C₉H₆F₆O
MW: 244.14 g/mol
Boiling Pt: 100 °C (14 mmHg)
Melting Pt: 55 °C
Flash Pt: 98 °C
MDL Number: MFCD00009907
CAS Number: 32707-89-4
EINECS: 251-168-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ef60aa1721220c0c2a086c63b56a8387fbe3d2aa&vid=ef60aa1721220c0c2a086c63b56a8387fbe3d2aa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR