1,2-Bis(2-iodoethoxy)ethane ≥96.0% stabilised

Supplier: TCI America

Synonyms: Ethylene Glycol Bis(2-iodoethyl) Ether

B1687-10G
TCB1687-010GEA 112.18 CAD
TCB1687-010G
1,2-Bis(2-iodoethoxy)ethane ≥96.0% stabilised
1,2-Bis(2-iodoethoxy)ethane

(stabilized with Copper chip)
CAS Number: 36839-55-1
MDL Number: MFCD00075008
Molecular Formula: C6H12I2O2
Molecular Weight: 369.97
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Flash Point (°C): 110
Specific Gravity (20/20): 2.05


Other Product Names / Synonyms: Ethylene Glycol Bis(2-iodoethyl) Ether

Formula: C₆H₁₂I₂O₂
MW: 369.97 g/mol
Density: 2.05 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00075008
CAS Number: 36839-55-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9467f98cd87c3058e07fd07aa39fd3ed5dafa463&vid=9467f98cd87c3058e07fd07aa39fd3ed5dafa463&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR