Benzyl bromoacetate ≥96.0%

Supplier: TCI America

Synonyms: Benzyl 2-bromoacetate, Bromoacetic benzyl ester

B1674-25G B1674-500G
TCB1674-025GEA 36.34 CAD
TCB1674-025G TCB1674-500G
Benzyl bromoacetate ≥96.0%
Benzyl bromoacetate

CAS Number: 5437-45-6
MDL Number: MFCD00000190
Molecular Formula: C9H9BrO2
Molecular Weight: 229.07
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 170
Flash Point (°C): 110
Specific Gravity (20/20): 1.46


Other Product Names / Synonyms: Bromoacetic Acid Benzyl Ester

Formula: C₉H₉BrO₂
MW: 229.07 g/mol
Boiling Pt: 170 °C
Density: 1.46 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00000190
CAS Number: 5437-45-6
EINECS: 226-611-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=906686bcc36586a91e64fc52c466dfa698b7a397&vid=906686bcc36586a91e64fc52c466dfa698b7a397&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR