5-Bromoisatin ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 5-Bromoindoline-2,3-dione, 5-Bromo-1H-indole-2,3-dione

B1566-250G B1566-25G
TCB1566-250GEA 1319.3 CAD
TCB1566-250G TCB1566-025G
5-Bromoisatin ≥97.0% (by HPLC, titration analysis)
5-Bromoisatin

CAS Number: 87-48-9
MDL Number: MFCD00005719
Molecular Formula: C8H4BrNO2
Molecular Weight: 226.03
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Melting point (°C): 253


Product Categories: Synthetic Organic Chemistry, Isatins (Multi-component Reaction)
Other Product Names / Synonyms: 5-Bromoindole-2,3-dione, 5-Bromo-2,3-indolinedione

Formula: C₈H₄BrNO₂
MW: 226.03 g/mol
Melting Pt: 253 °C
MDL Number: MFCD00149345
CAS Number: 87-48-9
EINECS: 201-747-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b35b5360c2372dc678552f003c9799aa01ac8882&vid=b35b5360c2372dc678552f003c9799aa01ac8882&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR