4-(2-Benzoxazolyl)-4'-(5-methyl-2-benzoxazolyl)stilbene ≥97.0% (by HPLC)

Supplier: TCI America

B1565-25G
TCB1565-25GEA 113.76 CAD
TCB1565-25G
4-(2-Benzoxazolyl)-4'-(5-methyl-2-benzoxazolyl)stilbene ≥97.0% (by HPLC)
4-(2-Benzoxazolyl)-4'-(5-methyl-2-benzoxazolyl)stilbene

CAS Number: 5242-49-9
Molecular Formula: C29H20N2O2
Molecular Weight: 428.49
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal

Formula: C₂₉H₂₀N₂O₂
CAS Number: 5242-49-9

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR