2,5-Bis(5-tert-butyl-2-benzoxazolyl)thiophene ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

B1554-5G B1554-25G
TCB1554-005GEA 58.46 CAD
TCB1554-005G TCB1554-025G
2,5-Bis(5-tert-butyl-2-benzoxazolyl)thiophene ≥98.0% (by HPLC, total nitrogen)
2,5-Bis(5-tert-butyl-2-benzoxazolyl)thiophene

CAS Number: 7128-64-5
MDL Number: MFCD00005774
Molecular Formula: C26H26N2O2S
Molecular Weight: 430.57
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 201
Lambda max.: 373 nm (EtOH)

Formula: C₂₆H₂₆N₂O₂S
Melting Pt: 201 °C
MDL Number: MFCD00005774
CAS Number: 7128-64-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4e2a0ca7aedb1de7cd6c69ec0b3715181eb5c055&vid=4e2a0ca7aedb1de7cd6c69ec0b3715181eb5c055&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR