4-Bromo-1,3-dimethoxybenzene ≥97.0%

Supplier: TCI America
Warning

B1481-25G B1481-5G
TCB1481-025GEA 198.29 CAD
TCB1481-025G 76219-344
4-Bromo-1,3-dimethoxybenzene ≥97.0%
4-Bromo-1,3-dimethoxybenzene

CAS Number: 17715-69-4
MDL Number: MFCD00009844
Molecular Formula: C8H9BrO2
Molecular Weight: 217.06
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 132
Flash Point (°C): 110
Freezing point (°C): 25

Formula: C₈H₉BrO₂
Boiling Pt: 132 °C
Density: 1.507 g/cm³ (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00009844
CAS Number: 17715-69-4
EINECS: 241-717-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1b228fc464f9aad37abe4629e4436360ee7dd63d&vid=1b228fc464f9aad37abe4629e4436360ee7dd63d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR