5-Bromo-2-chlorobenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

B1344-25G B1344-10G
TCB1344-025GEA 128.93 CAD
TCB1344-025G TCB1344-010G
5-Bromo-2-chlorobenzoic acid ≥98.0% (by GC, titration analysis)
5-Bromo-2-chlorobenzoic acid

CAS Number: 21739-92-4
MDL Number: MFCD00002415
Molecular Formula: C7H4BrClO2
Molecular Weight: 235.46
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 158

Formula: C₇H₄BrClO₂
Boiling Pt: 316 °C (1013 hPa)
Melting Pt: 158 °C
Storage Temperature: Ambient
MDL Number: MFCD00002415
CAS Number: 21739-92-4
EINECS: 244-558-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1d040cb8f68781ad9c8264bd2a04f9897a0dccda&vid=96229cf3e83c24da7655c19c7e168d462df2015f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR