(S)-(+)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: (S)-(+)-1,1'-Binaphthalene-2,2'-diyl-phosphate, (11bS)-4-Hydroxydinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide

B1144-1G B1144-100MG
TCB1144-001GEA 85.32 CAD
TCB1144-001G TCB1144-100MG
(S)-(+)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate ≥97.0% (by titrimetric analysis)
(S)-(+)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate

CAS Number: 35193-64-7
MDL Number: MFCD00010045
Molecular Formula: C20H13O4P
Molecular Weight: 348.29
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Specific rotation [a]20/D: 595 deg (C=1, MeOH)


Product Categories: Synthetic Organic Chemistry, Asymmetric Organocatalysts, Optical Resolving Agents, Analytical Science, e.e. / Absolute Configuration Determination (NMR)
Other Product Names / Synonyms: (S)-(+)-2,2'-Dihydroxy-1,1'-binaphthyl Hydrogen Phosphate

Formula: C₂₀H₁₃O₄P
MW: 348.289 g/mol
MDL Number: MFCD00010045
CAS Number: 35193-64-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=77550be6d972a553f390c960e3b4f08fd32f8a9d&vid=77550be6d972a553f390c960e3b4f08fd32f8a9d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR