4-Methyl-2-phenyl-1,3-dioxolane (mixture of isomers) ≥98.0%

Supplier: TCI America

Synonyms: Benzaldehyde Propylene Glycol Acetal

B1094-25ML
TCB1094-025MLEA 58.46 CAD
TCB1094-025ML
4-Methyl-2-phenyl-1,3-dioxolane (mixture of isomers) ≥98.0%
4-Methyl-2-phenyl-1,3-dioxolane (mixture of isomers)

CAS Number: 2568-25-4
MDL Number: MFCD00059732
Molecular Formula: C10H12O2
Molecular Weight: 164.20
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 125
Flash Point (°C): 113
Specific Gravity (20/20): 1.07


Other Product Names / Synonyms: Benzaldehyde Propylene Glycol Acetal

Formula: C₁₀H₁₂O₂
MW: 164.20 g/mol
Boiling Pt: 125 °C
Density: 1.07 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00059732
CAS Number: 2568-25-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b1ab6e5f45e635cd8c08a75266389c3f5d1d2208&vid=b1ab6e5f45e635cd8c08a75266389c3f5d1d2208&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR