Bis-trispropane ≥99.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 1,3-Bis(tris(hydroxymethyl)methylamino)propane, 1,3-Bis[tris(hydroxymethyl)amino]propane, 2,2'-(Propane-1,3-diylbis(azanediyl))bis(2-(hydroxymethyl)propane-1,3-diol), BIS-TRIS propane, 2,2'-(Propane-1,3-diyldiimino)bis[2-(hydroxymethyl)propane-1,3-diol]

B1057-25G
TCB1057-025GEA 144.93 CAD
TCB1057-025G
Bis-trispropane ≥99.0% (by titrimetric analysis)
BIS-TRIS propane

[for Buffer Material]
CAS Number: 64431-96-5
MDL Number: MFCD00004689
Molecular Formula: C11H26N2O6
Molecular Weight: 282.34
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 166


Product Categories: Bioscience, Buffer-related Reagents
Other Product Names / Synonyms: BIS-TRIS propane

Formula: C₁₁H₂₆N₂O₆
MW: 282.34 g/mol
Melting Pt: 166 °C
MDL Number: MFCD00004689
CAS Number: 64431-96-5
EINECS: 264-899-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=777a67054c3ade11154c36fbcecdaa8115f3a2ee&vid=c5a61813af44a22d1eb4a32c0b8e7379f28a5546&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR