2-Ethoxyethyl benzoate ≥98.0%

Supplier: TCI America

Synonyms: Ethylene Glycol Monoethyl Ether Benzoate, Benzoic Acid 2-Ethoxyethyl Ester, Cellosolve Benzoate

B0955-5ML
TCB0955-005MLEA 47.4 CAD
TCB0955-005ML
2-Ethoxyethyl benzoate ≥98.0%
2-Ethoxyethyl benzoate

CAS Number: 5451-72-9
MDL Number: MFCD00059371
Molecular Formula: C11H14O3
Molecular Weight: 194.23
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 261
Specific Gravity (20/20): 1.06


Other Product Names / Synonyms: Benzoic Acid 2-Ethoxyethyl Ester, Ethylene Glycol Monoethyl Ether Benzoate, Cellosolve Benzoate

Formula: C₁₁H₁₄O₃
MW: 194.23 g/mol
Boiling Pt: 261 °C
Density: 1.06 (20 °C)
MDL Number: MFCD00059371
CAS Number: 5451-72-9

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR