5-Bromosalicylaldehyde ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B0893-25G
TCB0893-500GEA 1790.14 CAD
TCB0893-500G TCB0893-025G
5-Bromosalicylaldehyde ≥98.0% (by GC, titration analysis)
5-Bromosalicylaldehyde

CAS Number: 1761-61-1
MDL Number: MFCD00003330
Molecular Formula: C7H5BrO2
Molecular Weight: 201.02
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 105


Other Product Names / Synonyms: 5-Bromo-2-hydroxybenzaldehyde

Formula: C₇H₅BrO₂
Melting Pt: 105 °C
Storage Temperature: Ambient
MDL Number: MFCD00003330
CAS Number: 1761-61-1
EINECS: 217-167-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4e9cd00dba9ba16b65585ce1c0e3cf8d5603aa92&vid=71bb4889a35f8807879b2929daf1c0e581f40261&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR