O-Benzyl-L-serine ≥99.0% (by titrimetric analysis)

Supplier: TCI America

B0861-1G B0861-5G
TCB0861-001GEA 50.56 CAD
TCB0861-001G TCB0861-005G
O-Benzyl-L-serine ≥99.0% (by titrimetric analysis)
O-Benzyl-L-serine

CAS Number: 4726-96-9
MDL Number: MFCD00065937
Molecular Formula: C10H13NO3
Molecular Weight: 195.22
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Specific rotation [a]20/D: 7.3 deg (C=2, 1mol/L HCl)


Product Categories: Bioscience, Side Chain Protected Amino Acids (N,C-termini Free)
Other Product Names / Synonyms: H-Ser(Bzl)-OH

Formula: C₁₀H₁₃NO₃
MDL Number: MFCD00065937
CAS Number: 4726-96-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1f31ff3f05aa457eedfd616f41a116b1a76fada6&vid=c416fc10e02cf014419c47e23d195a364eceaff4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR