2,2'-Dihydroxybiphenyl ≥98.0%

Supplier: TCI America

B0813-500G B0813-25G B0813-100G
TCB0813-500GEA 1083.88 CAD
TCB0813-500G TCB0813-025G TCB0813-100G
2,2'-Dihydroxybiphenyl ≥98.0%
2,2'-Dihydroxybiphenyl

CAS Number: 1806-29-7
MDL Number: MFCD00002210
Molecular Formula: C12H10O2
Molecular Weight: 186.21
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 325
Melting point (°C): 107
Flash Point (°C): 154


Product Categories: Materials Chemistry, Biphenyls (for High-Performance Polymer Research), Pressure & Heat Sensitive Recording Materials
Other Product Names / Synonyms: Biphenyl-2,2'-diol, 2,2'-Biphenol

Formula: C₁₂H₁₀O₂
Boiling Pt: 325 °C
Melting Pt: 107 °C
Density: 1.342 g/cm³ (20 °C)
Flash Pt: 154 °C
MDL Number: MFCD00002210
CAS Number: 1806-29-7
EINECS: 217-303-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b7892850df32fc743825b01a57b463affe4f9582&vid=b7892850df32fc743825b01a57b463affe4f9582&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR