Butyrophenone ≥99.0%

Supplier: TCI America

Synonyms: 1-Phenylbutan-1-one, Phenyl propyl ketone, 1-Phenyl-1-butanone

B0769-25ML B0769-500ML B0769-100ML
TCB0769-025MLEA 43.37 CAD
TCB0769-025ML TCB0769-500ML TCB0769-100ML
Butyrophenone ≥99.0%
Butyrophenone

CAS Number: 495-40-9
MDL Number: MFCD00009397
Molecular Formula: C10H12O
Molecular Weight: 148.21
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 221
Melting point (°C): 12
Flash Point (°C): 88
Specific Gravity (20/20): 0.99


Other Product Names / Synonyms: Phenyl Propyl Ketone

Formula: C₆H₅C(O)(CH₂)₂CH₃
MW: 148.2 g/mol
Boiling Pt: 221 °C
Melting Pt: 12 °C
Density: 0.99 (20 °C)
Flash Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD00009397
CAS Number: 495-40-9
EINECS: 207-799-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=480a64492a3195f352d5b0f17d550c62fc4db434&vid=2909b9c529d4e49a9bfc9def1a1dacb2cb6000d7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR