Trioctadecyl borate ≥97.0% (by titrimetric analysis)

Supplier: TCI America

B0523-25G
TCB0523-025GEA 199.08 CAD
TCB0523-025G
Trioctadecyl borate ≥97.0% (by titrimetric analysis)
Trioctadecyl borate

CAS Number: 5337-41-7
MDL Number: MFCD00026677
Molecular Formula: C54H111BO3
Molecular Weight: 819.29
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Boiling point (°C): 343
Melting point (°C): 53


Other Product Names / Synonyms: Boric Acid Trioctadecyl Ester, Boric Acid Tristearyl Ester, Tristearyl Borate

Formula: C₅₄H₁₁₁BO₃
Boiling Pt: 343 °C
Melting Pt: 53 °C
MDL Number: MFCD00026677
CAS Number: 5337-41-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18766f0193a99e868040e2183be695106ce2bc80&vid=18766f0193a99e868040e2183be695106ce2bc80&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR