Bindschedler's green leuco base ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

B0482-1G B0482-5G
TCB0482-001GEA 388.68 CAD
TCB0482-001G TCB0482-5G
Bindschedler's green leuco base ≥98.0% (by HPLC, titration analysis)
Bindschedler's green leuco base

CAS Number: 637-31-0
MDL Number: MFCD00059291
Molecular Formula: C16H21N3
Molecular Weight: 255.37
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 121


Other Product Names / Synonyms: Bis(4-dimethylaminophenyl)amine, Leuco Bindschedler's Green, N,N,N',N'-Tetramethyldiaminodiphenylamine

Formula: C₁₆H₂₁N₃
Melting Pt: 121 °C
MDL Number: MFCD00059291
CAS Number: 637-31-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d6b481414668e9d92ef064ee4e7f0d75aa5c685&vid=8d6b481414668e9d92ef064ee4e7f0d75aa5c685&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR