2-Phenylaniline ≥99.0% (by GC, titration analysis)

Supplier: TCI America
Warning

B0467-25G B0467-500G B0467-100G
TCB0467-025GEA 189.76 CAD
TCB0467-025G TCB0467-500G TCB0467-100G
2-Phenylaniline ≥99.0% (by GC, titration analysis)
2-Phenylaniline

[for Sugar determination]
CAS Number: 90-41-5
MDL Number: MFCD00007701
Molecular Formula: C12H11N
Molecular Weight: 169.23
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Boiling point (°C): 299
Melting point (°C): 51
Flash Point (°C): 153


Other Product Names / Synonyms: 2-Aminodiphenyl, 2-Biphenylamine

Formula: C₁₂H₁₁N
Boiling Pt: 299 °C
Melting Pt: 51 °C
Flash Pt: 153 °C
Storage Temperature: Ambient
MDL Number: MFCD00007701
CAS Number: 90-41-5
EINECS: 201-990-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36001bbae749308493888281673a7d5d7ad5ed64&vid=617abc533a597c58031d5564a82850379c7355d1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR