Benzyltriethylammonium iodide ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

B0446-25G B0446-5G
TCB0446-025GEA 203.35 CAD
TCB0446-025G TCB0446-5G
Benzyltriethylammonium iodide ≥98.0% (by titrimetric analysis)
Benzyltriethylammonium iodide

CAS Number: 5400-94-2
MDL Number: MFCD00043178
Molecular Formula: C13H22IN
Molecular Weight: 319.23
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 171

Formula: C₁₃H₂₂IN
Melting Pt: 171 °C
MDL Number: MFCD00043178
CAS Number: 5400-94-2
EINECS: 226-446-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e29775a4496995a475db98666d4c5076981e0b1d&vid=e29775a4496995a475db98666d4c5076981e0b1d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR