1-Butanesulfonic acid sodium salt ≥98.0% (by titrimetric analysis)

Supplier: TCI America

B0379-25G B0379-500G
TCB0379-025GEA 186.44 CAD
TCB0379-025G TCB0379-500G
1-Butanesulfonic acid sodium salt ≥98.0% (by titrimetric analysis)
1-Butansulphonic acid sodium salt

CAS Number: 2386-54-1
MDL Number: MFCD00007540
Molecular Formula: C4H10O3S
Molecular Weight: 160.16
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Other Product Names / Synonyms: 1-Butanesulfonic Acid Sodium Salt, Butylsulfonic Acid Sodium Salt

Formula: C₄H₉NaO₃S
Melting Pt: > 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007540
CAS Number: 2386-54-1
EINECS: 219-201-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=76d9ed426bfb187d352cd2da24404f6ddfd9fa42&vid=76d9ed426bfb187d352cd2da24404f6ddfd9fa42&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR