N-(4-Methoxy-2-hydroxybenzylidene)-4-butylaniline ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: N-(2-Hydroxy-p-anisal)-4-butylaniline, N-(2-Hydroxy-p-anisylidene)-4-butylaniline

B0250-1G
TCB0250-001GEA 464.52 CAD
TCB0250-001G
N-(4-Methoxy-2-hydroxybenzylidene)-4-butylaniline ≥98.0% (by HPLC, titration analysis)
N-(4-Methoxy-2-hydroxybenzylidene)-4-butylaniline

CAS Number: 30633-94-4
MDL Number: MFCD00059253
Molecular Formula: C18H21NO2
Molecular Weight: 283.37
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal


Product Categories: Materials Chemistry, Liquid Crystal Materials (Optically Functional Materials)
Other Product Names / Synonyms: N-(2-Hydroxy-p-anisal)-4-butylaniline, N-(2-Hydroxy-p-anisylidene)-4-butylaniline

Formula: C₁₈H₂₁NO₂
MW: 283.37 g/mol
MDL Number: MFCD00059253
CAS Number: 30633-94-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0761d9fc180e0f0545371a43dcc674d15d2786a3&vid=0761d9fc180e0f0545371a43dcc674d15d2786a3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR