1,4-Benzoquinone dioxime ≥95.0% (by total nitrogen basis)

Supplier: TCI America

Synonyms: p-Quinone Dioxime

B0090-25G
TCB0090-025GEA 34.76 CAD
TCB0090-025G
1,4-Benzoquinone dioxime ≥95.0% (by total nitrogen basis)
1,4-Benzoquinone dioxime

CAS Number: 105-11-3
MDL Number: MFCD00063636
Molecular Formula: C6H6N2O2
Molecular Weight: 138.13
Purity/Analysis Method: >95.0% (N)
Form: Crystal
Flash Point (°C): 184


Other Product Names / Synonyms: p-Quinone Dioxime

Formula: C₆H₆N₂O₂
MW: 138.13 g/mol
Flash Pt: 184 °C
MDL Number: MFCD00063636
CAS Number: 105-11-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0b44e0315654d57c6974e3705c417faa4d3df46f&vid=0b44e0315654d57c6974e3705c417faa4d3df46f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR