p-Benzoquinone ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: p-Quinone, Quinone, 1,4-Cyclohexadiene-3,6-dione, 1,4-Benzoquinone

B0089-25G B0089-100G
TCB0089-25GEA 50.69 CAD
TCB0089-25G TCB0089-100G
p-Benzoquinone ≥98.0% (by titrimetric analysis)
p-Benzoquinone

CAS Number: 106-51-4
MDL Number: MFCD00001591
Molecular Formula: C6H4O2
Molecular Weight: 108.10
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 113
Flash Point (°C): 78


Product Categories: Synthetic Organic Chemistry, C-H Bond Activation Reaction, Materials Chemistry, Molecular Conductors
Other Product Names / Synonyms: p-Quinone, p-Benzoquinone

Formula: C₆H₄O₂
MW: 108.1 g/mol
Boiling Pt: 174
Melting Pt: 113 °C
Density: 1.155 g/cm³ (20 °C)
Flash Pt: 78 °C
MDL Number: MFCD00000632
CAS Number: 106-51-4
EINECS: 214-028-8
UN: 2587
ADR: 6.1 II
Merck Index: 12,08259
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e5277bbbb6195cf354c2caf8f3aede75971c4933&vid=e5277bbbb6195cf354c2caf8f3aede75971c4933&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR