5-Acetyl-2-amino-4-methylthiazole ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

A2887-1G A2887-5G
TCA2887-1GEA 61.62 CAD
TCA2887-1G TCA2887-5G
5-Acetyl-2-amino-4-methylthiazole ≥98.0% (by GC, titration analysis)
5-Acetyl-2-amino-4-methylthiazole
Formula: C₆H₈N₂OS
Melting Pt: 266°C
MDL Number: MFCD00051952
CAS Number: 30748-47-1

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR