trans-4-Acetoxycinnamic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

A2845-25G A2845-5G
TCA2845-25GEA 799.48 CAD
TCA2845-25G TCA2845-5G
trans-4-Acetoxycinnamic acid ≥98.0% (by GC, titration analysis)
trans-4-Acetoxycinnamic acid
Formula: C₁₁H₁₀O₄
MDL Number: MFCD00016847
CAS Number: 27542-85-4
EINECS: 239-512-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 208.0 to 212.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR