3-Acetyl-6-bromo-2H-chromen-2-one ≥96.0% (by GC)

Supplier: TCI America
Warning

A2723-1G A2723-5G
TCA2723-1GEA 127.35 CAD
TCA2723-1G TCA2723-5G
3-Acetyl-6-bromo-2H-chromen-2-one ≥96.0% (by GC)
3-Acetyl-6-bromo-2H-chromen-2-one
Formula: C₁₁H₇BrO₃
MDL Number: MFCD00024075
CAS Number: 2199-93-1

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Green powder to crystal
Purity (GC) min. 96.0 %
Melting point 223.0 to 227.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR