3-Amino-4-(isobutylamino)quinoline hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: N4-Isobutylquinoline-3,4-diamine hydrochloride, 3-Amino-4-(isobutylamino)quinoline Hydrochloride

A2721-5G
TCA2721-1GEA 69.98 CAD
TCA2721-1G TCA2721-5G
3-Amino-4-(isobutylamino)quinoline hydrochloride ≥98.0% (by HPLC, titration analysis)
3-Amino-4-(isobutylamino)quinoline hydrochloride

CAS Number: 935521-01-0
MDL Number: MFCD08063180
Molecular Formula: C13H17N3
Molecular Weight: 251.76
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 264

Formula: C₁₃H₁₇N₃
MW: 251.76 g/mol
Melting Pt: 264 °C
MDL Number: MFCD08063180
CAS Number: 935521-01-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6ccc2b4aaa37c5e0454b8cac798f57e04dee2dfc&vid=6ccc2b4aaa37c5e0454b8cac798f57e04dee2dfc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR