4-Aminotriphenylamine ≥95.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

A2624-200MG
TCA2624-200MGEA 208.24 CAD
TCA2624-200MG
4-Aminotriphenylamine ≥95.0% (by HPLC, titration analysis)
4-Aminotriphenylamine
Formula: C₁₈H₁₆N₂
Boiling Pt: 270°C
MDL Number: MFCD00145130
CAS Number: 2350-01-8

Order Now

Specification Test Results

Appearance White to Brown powder to crystalline
Purity (HPLC) min. 95.0 area%
Purity (Nonaqueous Titration) min. 95.0 %
Melting point 144.0 to 148.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR