4-Allyloxy-2-hydroxybenzophenone ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

A2342-25G
TCA2342-25GEA 94.8 CAD
TCA2342-25G
4-Allyloxy-2-hydroxybenzophenone ≥98.0% (by HPLC, titration analysis)
4-Allyloxy-2-hydroxybenzophenone

CAS Number: 2549-87-3
MDL Number: MFCD00191713
Molecular Formula: C16H14O3
Molecular Weight: 254.29
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Boiling point (°C): 178
Melting point (°C): 68

Formula: C₁₆H₁₄O₃
MW: 254.2806 g/mol
Boiling Pt: 178 °C
Melting Pt: 68 °C
MDL Number: MFCD00191713
CAS Number: 2549-87-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=77db3587f0640fa8863112950426577574f5982c&vid=77db3587f0640fa8863112950426577574f5982c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR