3A-Amino-3A-deoxy-(2AS,3AS)-ɑ-cyclodextrin hydrate ≥90.0% (by HPLC)

Supplier: TCI America

Synonyms: 3A-Amino-3A-deoxy-(2AS,3AS)-α-cyclodextrin Hydrate

A2122-1G A2122-200MG
TCA2122-1GEA 502.11 CAD
TCA2122-1G TCA2122-200MG
3A-Amino-3A-deoxy-(2AS,3AS)-ɑ-cyclodextrin hydrate ≥90.0% (by HPLC)
3A-Amino-3A-deoxy-(2AS,3AS)-alpha-cyclodextrin hydrate

CAS Number: 121916-94-7
Molecular Formula: C36H61NO29
Molecular Weight: 971.86
Purity/Analysis Method: >90.0% (HPLC)
Form: Crystal
Specific rotation [a]20/D: 132.5 deg (C=1, H2O)

Formula: C₃₆H₆₁NO₂₉
MW: 971.86 g/mol
CAS Number: 121916-94-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=10d91c36c704665cc4a81ff04de7a8023e40cf4e&vid=10d91c36c704665cc4a81ff04de7a8023e40cf4e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (HPLC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR