4-Anilinopyridine ≥99.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: N-(4-Pyridyl)aniline, N-Phenylpyridin-4-amine

A2087-5G
TCA2087-5GEA 164.32 CAD
TCA2087-5G
4-Anilinopyridine ≥99.0% (by titrimetric analysis)
4-Anilinopyridine

CAS Number: 22961-45-1
MDL Number: MFCD00457758
Molecular Formula: C11H10N2
Molecular Weight: 170.22
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 177


Other Product Names / Synonyms: N-(4-Pyridyl)aniline

Formula: C₁₁H₁₀N₂
MW: 170.22 g/mol
Melting Pt: 177 °C
MDL Number: MFCD00457758
CAS Number: 22961-45-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f1d4b45b89b3c9dc26feb42ed0d3741cf1080be2&vid=f1d4b45b89b3c9dc26feb42ed0d3741cf1080be2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR