N-Acetyl-L-methionine ≥98.0%

Supplier: TCI America

Synonyms: N-Ac-Met-OH, N-Acetyl-L-Methionine, Ac-Met-OH NAM, Ac-Met-OH

A2056-25G A2056-5G
TCA2056-25GEA 54.65 CAD
TCA2056-25G TCA2056-5G
N-Acetyl-L-methionine ≥98.0%
N-Acetyl-L-methionine

CAS Number: 65-82-7
MDL Number: MFCD00064441
Molecular Formula: C7H13NO3S
Molecular Weight: 191.25
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 105
Specific rotation [a]20/D: -21 deg (C=1, H2O)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Ac-Met-OH

Formula: C₇H₁₃NO₃S
Melting Pt: 105 °C
MDL Number: MFCD00064441
CAS Number: 65-82-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce7d0fa7b1c0ff2b7f775990c3de4b3f12414301&vid=ce7d0fa7b1c0ff2b7f775990c3de4b3f12414301&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR