2-Amino-6-fluorobenzothiazole ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 6-Fluorobenzothiazol-2-ylamine, 6-Fluorobenzo[d]thiazol-2-amine, 6-Fluoro-1,3-benzothiazol-2-amine

A1993-5G
TCA1993-5GEA 124.82 CAD
TCA1993-5G
2-Amino-6-fluorobenzothiazole ≥97.0% (by HPLC, titration analysis)
2-Amino-6-fluorobenzothiazole

CAS Number: 348-40-3
MDL Number: MFCD00013336
Molecular Formula: C7H5FN2S
Molecular Weight: 168.19
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Melting point (°C): 183

Formula: C₇H₅FN₂S
MW: 168.19 g/mol
Boiling Pt: 304
Melting Pt: 183 °C
MDL Number: MFCD00013336
CAS Number: 348-40-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9aecd2bfda90eca8662618fae8261608aaa9b7f&vid=e9aecd2bfda90eca8662618fae8261608aaa9b7f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR