2-Amino-5-nitro-3-picoline ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A1891-5G A1891-1G
TCA1891-5GEA 120.08 CAD
TCA1891-5G TCA1891-1G
2-Amino-5-nitro-3-picoline ≥98.0% (by GC, titration analysis)
2-Amino-5-nitro-3-picoline

CAS Number: 18344-51-9
Molecular Formula: C6H7N3O2
Molecular Weight: 153.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 261
Melting point (°C): 255


Other Product Names / Synonyms: 2-Amino-5-nitro-3-picoline

Formula: C₆H₇N₃O₂
Boiling Pt: 261 °C
Melting Pt: 255 °C
MDL Number: MFCD03095060
CAS Number: 18344-51-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6544f3d1ef5891973ba2acdcf15a070714a0cfe6&vid=6544f3d1ef5891973ba2acdcf15a070714a0cfe6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR