N-Acetylneuraminic acid methyl ester ≥95.0% (by HPLC)

Supplier: TCI America

Synonyms: Methyl 5-Acetamido-3,5-dideoxy-D-glycero-D-galacto-2-nonulopyranosonate, (2S,4S,5R,6R)-Methyl 5-acetamido-2,4-dihydroxy-6-((1R,2R)-1,2,3-trihydroxypropyl)tetrahydro-2H-pyran-2-carboxylate

A1821-1G
TCA1821-1GEA 221.52 CAD
TCA1821-1G
N-Acetylneuraminic acid methyl ester ≥95.0% (by HPLC)
N-Acetylneuraminic acid methyl ester

CAS Number: 22900-11-4
Molecular Formula: C12H21NO9
Molecular Weight: 323.30
Purity/Analysis Method: >95.0% (HPLC)
Form: Crystal
Flash Point (°C): 113
Specific rotation [a]20/D: -27.5 deg (C=1, H2O)
Storage Temperature: <0°C


Product Categories: Bioscience, Sialic Acids
Other Product Names / Synonyms: Methyl 5-Acetamido-3,5-dideoxy-D-glycero-D-galacto-2-nonulopyranosonate

Formula: C₁₂H₂₁NO₉
MW: 323.30 g/mol
Flash Pt: 113 °C
MDL Number: MFCD08276447
CAS Number: 22900-11-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea460c7965a8329f6cc393963b93ae6a71d650a3&vid=ea460c7965a8329f6cc393963b93ae6a71d650a3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR