3-Aminophenylboronic acid hemisulfate ≥98%

Supplier: TCI America

A1774-5G A1774-1G A1774-25G
TCA1774-5GEA 150.1 CAD
TCA1774-5G TCA1774-1G TCA1774-25G
3-Aminophenylboronic acid hemisulfate ≥98%
3-Aminophenylboronic acid hemisulphate

CAS Number: 66472-86-4
Molecular Formula: C6H8BNO2
Molecular Weight: 185.98
Form: Crystal
Color: White


Other Product Names / Synonyms: 3-Aminobenzeneboronic Acid Hemisulfate

Formula: C₁₂H₁₈B₂N₂O₈S
MDL Number: MFCD00013111
CAS Number: 66472-86-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3281a0c39a4ef9055fc60dfc8a986eabb5e3c699&vid=3281a0c39a4ef9055fc60dfc8a986eabb5e3c699&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR