1-Adamantyl isocyanate ≥98.0% (by GC)

Supplier: TCI America
Warning

A1706-1G A1706-5G
TCA1706-1GEA 292.3 CAD
TCA1706-1G TCA1706-5G
1-Adamantyl isocyanate ≥98.0% (by GC)
1-Adamantyl isocyanate
Formula: C₁₁H₁₅NO
MDL Number: MFCD00134405
CAS Number: 4411-25-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 145.0 to 149.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR