2-Amino-4-methylbenzothiazole ≥98.0% (by titrimetric analysis)

Supplier: TCI America

A1344-25G
TCA1344-25GEA 146.94 CAD
TCA1344-25G
2-Amino-4-methylbenzothiazole ≥98.0% (by titrimetric analysis)
2-Amino-4-methylbenzothiazole

CAS Number: 1477-42-5
MDL Number: MFCD00005793
Molecular Formula: C8H8N2S
Molecular Weight: 164.23
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 137

Formula: C₈H₈N₂S
Melting Pt: 137 °C
MDL Number: MFCD00005793
CAS Number: 1477-42-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2d9523009cba73db7fad7192c71b019ac95a71b6&vid=2d9523009cba73db7fad7192c71b019ac95a71b6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR