3-Pyridyl acetate ≥98.0%

Supplier: TCI America

Synonyms: Pyridin-3-yl acetate, 3-Acetoxypyridine, 3-Pyridyl acetate

A1210-25G
TCA1210-025GEA 189.6 CAD
TCA1210-025G
3-Pyridyl acetate ≥98.0%
3-Pyridyl acetate

CAS Number: 17747-43-2
MDL Number: MFCD00006377
Molecular Formula: C7H7NO2
Molecular Weight: 137.14
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 137
Flash Point (°C): 100
Specific Gravity (20/20): 1.15


Other Product Names / Synonyms: 3-Pyridyl Acetate

Formula: C₇H₇NO₂
MW: 137.14 g/mol
Boiling Pt: 137 °C
Density: 1.15 (20 °C)
Flash Pt: 100 °C
MDL Number: MFCD00006377
CAS Number: 17747-43-2
EINECS: 241-742-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cd839a084e5df1516935145f16fada4bb63b849&vid=9cd839a084e5df1516935145f16fada4bb63b849&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR