Tributyl acetylcitrate ≥97.0%

Supplier: TCI America

A0822-25ML A0822-500ML
TCA0822-025MLEA 28.88 CAD
TCA0822-025ML TCA0822-500ML
Tributyl acetylcitrate ≥97.0%
Tributyl acetylcitrate

CAS Number: 77-90-7
MDL Number: MFCD00043554
Molecular Formula: C20H34O8
Molecular Weight: 402.48
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 343
Melting point (°C): -80
Flash Point (°C): 204
Specific Gravity (20/20): 1.05


Product Categories: Materials Chemistry, Resin Additives
Other Product Names / Synonyms: O-Acetylcitric Acid Tributyl Ester

Formula: C₂₀H₃₄O₈
Boiling Pt: 343 °C
Melting Pt: –80 °C
Density: 1.05 (20 °C)
Flash Pt: 204 °C
MDL Number: MFCD00043554
CAS Number: 77-90-7
EINECS: 201-067-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3feebdcf5a9ee49df987457165cf17f21f9d6672&vid=3feebdcf5a9ee49df987457165cf17f21f9d6672&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR