Nɑ-Acetyl-L-glutamine ≥99.0% (by titrimetric analysis)

Supplier: TCI America

A0784-25G
TCA0784-025GEA 52.14 CAD
TCA0784-025G
Nɑ-Acetyl-L-glutamine ≥99.0% (by titrimetric analysis)
Nalpha-Acetyl-L-glutamine

CAS Number: 2490-97-3
MDL Number: MFCD00038159
Molecular Formula: C7H12N2O4
Molecular Weight: 188.18
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 208
Specific rotation [a]20/D: -12 deg (C=3, H2O)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Ac-Gln-OH

Formula: C₇H₁₂N₂O₄
Melting Pt: 208 °C
MDL Number: MFCD00038159
CAS Number: 2490-97-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a5327e9cc73488dacd95af2a2fe2541fe3362160&vid=a5327e9cc73488dacd95af2a2fe2541fe3362160&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR