3-Aminocyclohexanecarboxylic acid (cis and trans mixture) ≥95.0% (by titrimetric analysis)

Supplier: TCI America

A0749-5G A0749-25G
TCA0749-005GEA 128.93 CAD
TCA0749-005G TCA0749-025G
3-Aminocyclohexanecarboxylic acid (cis and trans mixture) ≥95.0% (by titrimetric analysis)
3-Aminocyclohexanecarboxylic acid (cis and trans mixture)

CAS Number: 25912-50-9
MDL Number: MFCD00059562
Molecular Formula: C7H13NO2
Molecular Weight: 143.19
Purity/Analysis Method: >95.0% (T)
Form: Crystal

Formula: C₇H₁₃NO₂
MDL Number: MFCD00059562
CAS Number: 25912-50-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48517bc66c9e6a999a73099613b229721effddb9&vid=48517bc66c9e6a999a73099613b229721effddb9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR