Azo Blue

Supplier: TCI America

A0587-500G A0587-25G
TCA0587-500GEA 520.61 CAD
TCA0587-500G TCA0587-025G
Azo Blue
Azo Blue

CAS Number: 6059-34-3
MDL Number: MFCD00059133
Molecular Formula: C34H26N4O8S2
Molecular Weight: 726.69
Form: Crystal
Color: Black


Other Product Names / Synonyms: Direct Violet 39

Formula: C₃₄H₂₆N₄O₈S₂
MDL Number: MFCD00059133
CAS Number: 6059-34-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d90154c7182792636d6caac1534c2202f9f6412c&vid=d90154c7182792636d6caac1534c2202f9f6412c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR