Mordant blue 13

Supplier: TCI America

A0584-25G
TCA0584-025GEA 140.62 CAD
TCA0584-025G
Mordant blue 13
Mordant blue 13

CAS Number: 1058-92-0
MDL Number: MFCD00003941
Molecular Formula: C16H11ClN2O9S2
Molecular Weight: 518.80
Form: Crystal
Color: Deep Yellow


Other Product Names / Synonyms: 3-(5-Chloro-2-hydroxyphenylazo)-4,5-dihydroxy-2,7-naphthalenedisulfonic Acid Disodium Salt, Eriochrome Blue SE, Sunchromine Fast Blue MB, Plasmocorinth B

Formula: C₁₆H₁₁ClN₂O₉S₂
MDL Number: MFCD00003941
CAS Number: 1058-92-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=edf42a4e10548f773570f430d6d7838aca88d415&vid=edf42a4e10548f773570f430d6d7838aca88d415&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR