6-O-Palmitoyl-L-ascorbic acid ≥97.0% (by titrimetric analysis)

Supplier: TCI America

A0540-25G
TCA0540-025GEA 36.34 CAD
TCA0540-025G
6-O-Palmitoyl-L-ascorbic acid ≥97.0% (by titrimetric analysis)
6-O-Palmitoyl-L-ascorbic acid

CAS Number: 137-66-6
MDL Number: MFCD00005377
Molecular Formula: C22H38O7
Molecular Weight: 414.54
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 114
Specific rotation [a]20/D: 22.5 deg (C=1, EtOH)


Product Categories: Bioscience, Sugar Acids, Sugars (Others), Vitamin Derivatives
Other Product Names / Synonyms: L-Ascorbyl 6-Palmitate

Formula: C₂₂H₃₈O₇
Melting Pt: 114 °C
MDL Number: MFCD00005377
CAS Number: 137-66-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c770731bde1c827beb187b6f64498c035d83ad13&vid=c770731bde1c827beb187b6f64498c035d83ad13&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR