3-Amino-1-propanol hydrochloride ≥95.0% (by titrimetric analysis)

Supplier: TCI America

A0407-25G
TCA0407-025GEA 60.04 CAD
TCA0407-025G
3-Amino-1-propanol hydrochloride ≥95.0% (by titrimetric analysis)
3-Amino-1-propanol hydrochloride

CAS Number: 14302-46-6
MDL Number: MFCD00060203
Molecular Formula: C3H9NO
Molecular Weight: 111.57
Purity/Analysis Method: >95.0% (T)
Form: Clear Liquid


Other Product Names / Synonyms: Propanolamine Hydrochloride

Formula: C₃H₉NO
MDL Number: MFCD00060203
CAS Number: 14302-46-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a83f848803c3db03067a5a3fb47efc9758ede731&vid=a83f848803c3db03067a5a3fb47efc9758ede731&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR