Cysteaminium chloride ≥95.0% (by titrimetric analysis)

Supplier: TCI America

A0296-100G
TCA0296-100GEA 175.38 CAD
TCA0296-100G
Cysteaminium chloride ≥95.0% (by titrimetric analysis)
Cysteaminium chloride

CAS Number: 156-57-0
MDL Number: MFCD00012904
Molecular Formula: C2H7NS
Molecular Weight: 113.60
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 68
Flash Point (°C): 66
Storage Temperature: 0-10°C


Product Categories: Bioscience, Reagents for Inflammation & Immunity Disorder Models, Antioxidants, Functionality Creating Reagents
Other Product Names / Synonyms: Cysteamine Hydrochloride, 2-Mercaptoethylamine Hydrochloride

Formula: C₂H₇NS·HCl
Melting Pt: 68 °C
Flash Pt: 66 °C
MDL Number: MFCD00012904
CAS Number: 156-57-0
EINECS: 205-858-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3cb100e0a19c16a9e07a4582e7c35400a544d73b&vid=3cb100e0a19c16a9e07a4582e7c35400a544d73b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR