4-Aminobutyric acid ≥99.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: GABA, γ-Aminobutyric acid

A0282-100G
TCA0282-100GEA 160.64 CAD
TCA0282-100G
4-Aminobutyric acid ≥99.0% (by titrimetric analysis)
4-Aminobutyric acid

CAS Number: 56-12-2
MDL Number: MFCD00008226
Molecular Formula: C4H9NO2
Molecular Weight: 103.12
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 195


Product Categories: Bioscience, Neurotransmitter Agent Ingredients, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: GABA

Formula: C₄H₉NO₂
MW: 103.12 g/mol
Boiling Pt: 248 °C (1013 hPa)
Melting Pt: 195 °C
Density: 1.21 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008226
CAS Number: 56-12-2
EINECS: 200-258-6
Merck Index: 13,00429
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ed10fbfb285efa2f0e0517a13ab3442baf4bbaa&vid=1ed10fbfb285efa2f0e0517a13ab3442baf4bbaa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR